Assay Details
| Gene Symbol: RRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042908.1 | - | 1 | 347 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | frequently rearranged in advanced T-cell lymphomas 2 | FRAT2 | -58.0kb | Sense | Chr. 10 - 97332497 - 97334701 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC644215 | LOC644215 | 7.2kb | Sense | Chr. 10 - 97401242 - 97426252 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RRP12
Genomic Map