This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03657708_m1
See other LOC339874 GE Assays ›
Gene Symbol:
LOC339874
Gene Name:
uncharacterized LOC339874
Gene Aliases:
-
Chromosome Location:
Chr.3: 131325092 - 131381475 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03657708_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC339874
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038976.1 3 - 4 797 73
GenBank mRNA BC043572.1 - 3 - 4 797 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NIMA related kinase 11 NEK11 Anti-sense Chr. 3 - 131026850 - 131350465 on NCBI Build GRCh38
Downstream nudix hydrolase 16 pseudogene 1 NUDT16P1 NUDT16P 34.1kb Anti-sense Chr. 3 - 131361845 - 131365122 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012440.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001786.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Agilent Affymetrix ArrayStar

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585523 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726371 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15489874 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726373 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585526 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726370 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726369 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585535 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726383 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726372 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585528 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726378 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585532 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726377 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726368 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15489880 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02585530 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726366 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726386 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726376 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22726389 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 111186 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 15973 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 889722 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 708540 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 162917 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 343316 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 576365 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 911874 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 173516 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 30223 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 213602 AF243385.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 213602 BC031058.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 213602 BC048215.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 213602 NM_006750.3 refseq_mrna_human_NM
Agilent SurePrint G3 Human CNV Microarray 2x400K 213602 U40572.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 14894 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 413157 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 291446 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 299890 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 259940 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3323979 - -
Agilent Human Genome CGH Microarray 44B A_14_P115035 - -
Agilent Human Genome CGH Microarray 44A A_14_P115035 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3239 - -
Agilent Human Genome CGH + SNP Microarray A_16_P36451834 - -
Agilent Human Genome CGH + SNP Microarray A_18_P23169793 - -
Agilent Human Genome CGH + SNP Microarray A_18_P23169793 - -
Agilent Human Genome CGH + SNP Microarray A_16_P58563729 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3323979 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3323979 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P36451834 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P115035 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P36451798 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P36451820 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P36451845 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00812124 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00812092 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P115035 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P36451834 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 35420 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 12548 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 68118 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 27613 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36451834 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36451820 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36451798 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00812089 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P115035 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16406263 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16406270 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P14390700 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00812092 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00812124 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00812107 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00812100 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36451795 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36451845 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36451834 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00812124 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36451845 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00812092 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36451820 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36451798 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1561348_PM_at - -
Affymetrix HT HG-U133+ PM Array Plate 1562638_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR03032325.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03032325.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032325.hg.1 TC03001786.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032317.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03032317.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032317.hg.1 TC03001786.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03032315.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03032315.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032315.hg.1 TC03001786.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03032326.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03032326.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032326.hg.1 TC03001786.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03032316.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03032316.hg.1 NR_038976.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03032316.hg.1 TC03001786.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000441345.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 ENST00000513905.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay TC0300012440.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193019.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193019.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193019.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193019.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193019.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193020.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193020.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193020.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193020.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193020.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193031.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193031.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193031.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193031.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193031.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193032.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193032.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193032.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193032.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193032.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193022.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 ENST00000511699.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193023.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193018.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193018.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193018.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193018.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193018.hg.1 TC0300012440.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300193017.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193017.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193017.hg.1 ENST00000502521.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300193017.hg.1 NR_038976.1 Refseq
Affymetrix Human Clariom D Assay PSR0300193017.hg.1 TC0300012440.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 1562638_at - -
Human Genome U133 Plus 2.0 Array 1562638_PM_at - -
Human Genome U133 Plus 2.0 Array 1561348_at - -
Human Genome U133 Plus 2.0 Array 1561348_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1561348_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562638_at9 - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A021601 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A021600 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A021600 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A021601 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline