This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03661175_m1
See other PSMG4 GE Assays ›
Gene Symbol:
PSMG4
Gene Name:
proteasome assembly chaperone 4
Gene Aliases:
C6orf86, PAC4, bA506K6.2
Chromosome Location:
Chr.6: 3254325 - 3308224 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03661175_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PSMG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125314.1 - 1 - 2 761 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC100507194 LOC100507194 -35.6kb Anti-sense Chr. 6 - 3182817 - 3195767 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014058.hg.1 ENST00000416079.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014058.hg.1 ENST00000416079.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014058.hg.1 ENST00000416079.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
Affymetrix Human Clariom D Assay TC0600014058.hg.1 ENST00000416079.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014058.hg.1 ENST00000416079.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014058.hg.1 ENST00000416079.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0