This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03662100_s1
See other ALDH18A1 GE Assays ›
Gene Symbol:
ALDH18A1
Gene Name:
aldehyde dehydrogenase 18 family member A1
Gene Aliases:
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
Chromosome Location:
Chr.10: 95605929 - 95656810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03662100_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH18A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126355.1 - 1 2648 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW966198.1 - 1 37 Chr. 10 - 95606689 - 95607161 [-] on Build GRCh38 78
GenBankmRNA AA337494.1 - 1 38 Chr. 10 - 95606868 - 95607162 [-] on Build GRCh38 78
GenBankmRNA BQ723247.1 - 1 606 Chr. 10 - 95606861 - 95607730 [-] on Build GRCh38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sorbin and SH3 domain containing 1 SORBS1 CAP,
FLAF2,
R85FL,
SH3D5,
SH3P12,
SORB1
-44.5kb Sense Chr. 10 - 95311773 - 95561420 on NCBI Build GRCh38
Downstream tectonic family member 3 TCTN3 C10orf61,
JBTS18,
OFD4,
TECT3
53.6kb Sense Chr. 10 - 95663396 - 95694143 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 ENST00000419777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 ENST00000419777.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 NR_121652.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 NR_121652.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007110.hg.1 NR_121652.1 Refseq
Affymetrix Human Clariom D Assay TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007110.hg.1 ENST00000412244.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007110.hg.1 ENST00000419777.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007110.hg.1 ENST00000419777.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007110.hg.1 NR_121652.1 Refseq
Affymetrix Human Clariom D Assay TC1000007110.hg.1 NR_121652.1 Refseq
Affymetrix Human Clariom D Assay TC1000007110.hg.1 NR_121652.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0