This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03669487_s1
See other DLGAP4 GE Assays ›
Gene Symbol:
DLGAP4
Gene Name:
DLG associated protein 4
Gene Aliases:
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4
Chromosome Location:
Chr.20: 36306323 - 36528637 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03669487_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLGAP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057672.1 - 1 1554 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB348037.1 - 1 Chr. 20 - 36470029 - 36470575 [-] on Build GRCh38 97
GenBankmRNA BM987094.1 - 1 Chr. 20 - 36470139 - 36470590 [-] on Build GRCh38 97
GenBankmRNA AA328499.1 - 1 217 Chr. 20 - 36470178 - 36470534 [+] on Build GRCh38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream AAR2 splicing factor homolog AAR2 C20orf4,
CGI-23
-211.9kb Sense Chr. 20 - 36236136 - 36256941 on NCBI Build GRCh38
Downstream DLGAP4 antisense RNA 1 DLGAP4-AS1 CCAT7 37.1kb Anti-sense Chr. 20 - 36507702 - 36573275 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline