This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03670033_s1
See other SLC9A5 GE Assays ›
Gene Symbol:
SLC9A5
Gene Name:
solute carrier family 9 member A5
Gene Aliases:
NHE5
Chromosome Location:
Chr.16: 67247549 - 67272204 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03670033_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC9A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK091748.1 - 1 753 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ773107.1 - 1 Chr. 16 - 67261488 - 67262217 [-] on Build GRCh38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream formin homology 2 domain containing 1 FHOD1 FHOS -13.6kb Anti-sense Chr. 16 - 67229389 - 67247522 on NCBI Build GRCh38
Downstream pleckstrin homology and RhoGEF domain containing G4 PLEKHG4 ARHGEF44,
PRTPHN1,
SCA4
14.0kb Sense Chr. 16 - 67277820 - 67289500 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011413.hg.1 ENST00000573063.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011413.hg.1 ENST00000573063.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011413.hg.1 ENST00000573063.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011413.hg.1 ENST00000573063.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0