This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03670619_s1
See other ATP5J2 GE Assays ›
Gene Symbol:
ATP5J2
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Gene Aliases:
ATP5JL
Chromosome Location:
Chr.7: 99458161 - 99466201 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03670619_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5J2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK094388.1 - 1 1455 68
AK123419.1 - 1 3118 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB297266.1 - 1 Chr. 7 - 99458190 - 99458742 [+] on Build GRCh38 68
GenBankmRNA BU680821.1 - 1 Chr. 7 - 99458195 - 99458905 [+] on Build GRCh38 68
GenBankmRNA BU688836.1 - 1 Chr. 7 - 99458195 - 99458673 [+] on Build GRCh38 68
GenBankmRNA AI985019.1 - 1 Chr. 7 - 99458190 - 99458624 [+] on Build GRCh38 68
GenBankmRNA BM980489.1 - 1 Chr. 7 - 99458195 - 99458942 [+] on Build GRCh38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATP5J2-PTCD1 readthrough ATP5J2-PTCD1 Sense Chr. 7 - 99416739 - 99466201 on NCBI Build GRCh38
Downstream zinc finger protein 789 ZNF789 13.0kb Anti-sense Chr. 7 - 99472819 - 99487600 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0