This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673222_s1
See other LOC283484 GE Assays ›
Gene Symbol:
LOC283484
Gene Name:
uncharacterized LOC283484
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673222_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC283484
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK127899.1 - 1 1948 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW665184.1 - 1 Chr. 13 - 107872883 - 107873372 [-] on Build GRCh38 101
GenBankmRNA AI203870.1 - 1 Chr. 13 - 107872878 - 107873364 [-] on Build GRCh38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream family with sequence similarity 155 member A FAM155A -4.0kb Anti-sense Chr. 13 - 107168531 - 107867255 on NCBI Build GRCh38
Downstream DNA ligase 4 LIG4 LIG4S 334.1kb Anti-sense Chr. 13 - 108207442 - 108218368 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569799 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569809 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569803 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P09120887 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569800 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569812 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569807 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569801 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569802 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P29569808 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P09120889 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P09120894 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 72105 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 902935 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 184777 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3679941 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49660 - -
Agilent Human Genome CGH + SNP Microarray A_16_P40091997 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3679941 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40091997 - -
Agilent Human 1B Microarray G4111A A_32_P467509 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P40091997 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1556940_PM_at - -
Affymetrix HT HG-U133+ PM Array Plate 1556941_PM_a_at - -
Human Genome U133 Plus 2.0 Array 1556941_a_at - -
Human Genome U133 Plus 2.0 Array 1556941_PM_a_at - -
Human Genome U133 Plus 2.0 Array 1556940_PM_at - -
Human Genome U133 Plus 2.0 Array 1556940_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556940_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1556941_a_at9 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline