This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673449_s1
See other ETFB GE Assays ›
Gene Symbol:
ETFB
Gene Name:
electron transfer flavoprotein beta subunit
Gene Aliases:
FP585, MADD
Chromosome Location:
Chr.19: 51345155 - 51366418 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673449_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETFB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK128845.1 - 1951 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000309244.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000354232.8 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000593992.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000594361.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 ENST00000596253.1 Ensembl
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001014763.1 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq
Affymetrix Human Clariom D Assay TC1900012016.hg.1 NM_001985.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0