This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673714_s1
See other APPBP2 GE Assays ›
Gene Symbol:
APPBP2
Gene Name:
amyloid beta precursor protein binding protein 2
Gene Aliases:
APP-BP2, HS.84084, PAT1
Chromosome Location:
Chr.17: 60443149 - 60526240 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673714_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APPBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL157428.1 - 2 2151 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein L12 pseudogene 38 RPL12P38 RPL12_14_1564 -25.2kb Sense Chr. 17 - 60433678 - 60434300 on NCBI Build GRCh38
Downstream uncharacterized LOC388406 LOC388406 101.9kb Anti-sense Chr. 17 - 60564546 - 60586620 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000083182.7 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000585368.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000588668.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000589341.5 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000590244.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 ENST00000592995.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_001282476.1 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq
Affymetrix Human Clariom D Assay TC1700011319.hg.1 NM_006380.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5vr65:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0