This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03674913_s1
See other NFE2L2 GE Assays ›
Gene Symbol:
NFE2L2
Gene Name:
nuclear factor, erythroid 2 like 2
Gene Aliases:
HEBP1, NRF2
Chromosome Location:
Chr.2: 177230303 - 177265131 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03674913_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFE2L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF323119.1 - 152 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF808384.1 - 1 Chr. 20 - 481799 - 482260 [+] on Build GRCh38 69
GenBankmRNA BU663805.1 - 1 396 Chr. 20 - 481868 - 482556 [-] on Build GRCh38 69
GenBankmRNA BG319490.1 - 1 Chr. 20 - 481991 - 482420 [+] on Build GRCh38 69
GenBankmRNA BQ890328.1 - 1 686 Chr. 20 - 481958 - 482872 [-] on Build GRCh38 69
GenBankmRNA BU934015.1 - 1 479 Chr. 20 - 481910 - 482662 [-] on Build GRCh38 69
GenBankmRNA BF740009.1 - 1 388 Chr. 20 - 482099 - 482546 [-] on Build GRCh38 69
GenBankmRNA BQ422912.1 - 1 516 Chr. 20 - 481859 - 482693 [-] on Build GRCh38 69
GenBankmRNA W26334.1 - 71 Chr. - - [-] on Build 69
GenBankmRNA BM558699.1 - 1 546 Chr. 20 - 481713 - 482732 [-] on Build GRCh38 69
GenBankmRNA CA313977.1 - 1 Chr. 20 - 481513 - 482329 [+] on Build GRCh38 69
GenBankmRNA BU742389.1 - 1 Chr. 20 - 481513 - 482278 [+] on Build GRCh38 69
GenBankmRNA BG110823.1 - 1 499 Chr. 20 - 482021 - 482675 [-] on Build GRCh38 69
GenBankmRNA CX785986.1 - 1 277 Chr. 20 - 481668 - 482463 [-] on Build GRCh38 69
GenBankmRNA BF748004.1 - 1 349 Chr. 20 - 482051 - 482472 [-] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TBC1 domain family member 20 TBC1D20 C20orf140,
WARBM4
-19.3kb Sense Chr. 20 - 435477 - 462553 on NCBI Build GRCh38
Downstream transcription factor 15 (basic helix-loop-helix) TCF15 EC2,
PARAXIS,
bHLHa40
121.4kb Sense Chr. 20 - 603993 - 610266 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF323119.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline