This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03675262_s1
See other COTL1 GE Assays ›
Gene Symbol:
COTL1
Gene Name:
coactosin like F-actin binding protein 1
Gene Aliases:
CLP
Chromosome Location:
Chr.16: 84565596 - 84618096 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03675262_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COTL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125645.1 - 1 1648 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TBC/LysM-associated domain containing 1 TLDC1 KIAA1609 -14.2kb Sense Chr. 16 - 84476355 - 84504752 on NCBI Build GRCh38
Downstream kelch like family member 36 KLHL36 C16orf44 77.3kb Anti-sense Chr. 16 - 84648504 - 84667686 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000262428.4 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000561707.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564057.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000564662.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567278.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 ENST00000567786.2 Ensembl
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq
Affymetrix Human Clariom D Assay TC1600011060.hg.1 NM_021149.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t6srt:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0