This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03678832_s1
See other SSR4P1 GE Assays ›
Gene Symbol:
SSR4P1
Gene Name:
signal sequence receptor subunit 4 pseudogene 1
Gene Aliases:
C21orf122, PRED57, PRED90
Chromosome Location:
Chr.21: 45070955 - 45073211 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03678832_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSR4P1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027292.2 1 649 85
GenBank mRNA BC004343.1 - 1 649 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000415143.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000415143.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000429427.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 ENST00000599569.2 Ensembl
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq
Affymetrix Human Clariom D Assay TC2100008404.hg.1 NR_027292.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline