This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03679320_m1
See other IGFALS GE Assays ›
Gene Symbol:
IGFALS
Gene Name:
insulin like growth factor binding protein acid labile subunit
Gene Aliases:
ACLSD, ALS
Chromosome Location:
Chr.16: 1790413 - 1794908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03679320_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFALS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027389.1 1 - 2 73 99
GenBank mRNA DA642067.1 - 1 - 2 73 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA637489.1 - 1 - 2 136 Chr. 16 - 1791986 - 1794971 [-] on Build GRCh38 99
GenBankmRNA DA641095.1 - 1 - 2 73 Chr. 16 - 1791934 - 1794908 [-] on Build GRCh38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nucleotide binding protein 2 NUBP2 CFD1,
NBP 2,
NUBP1
-1.2kb Anti-sense Chr. 16 - 1782923 - 1789191 on NCBI Build GRCh38
Downstream hydroxyacylglutathione hydrolase HAGH GLO2,
GLX2,
GLXII,
HAGH1
14.2kb Sense Chr. 16 - 1809099 - 1827194 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011461.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000761.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
signal transduction
cellular protein metabolic process
insulin-like growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 ENST00000215539.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 ENST00000415638.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011124.hg.1 TC16000761.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16011127.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011127.hg.1 TC16000761.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600192186.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600192186.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192186.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600192186.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192186.hg.1 TC1600011461.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192180.hg.1 TC1600011461.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0