This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03680785_g1
See other LINC00998 GE Assays ›
Gene Symbol:
LINC00998
Gene Name:
long intergenic non-protein coding RNA 998
Gene Aliases:
-
Chromosome Location:
Chr.7: 113116718 - 113118582 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03680785_g1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00998
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024412.1 1 - 2 142 95
GenBank mRNA DN995392.1 - 1 - 2 116 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN430142.1 - 1 - 2 101 Chr. 7 - 113117303 - 113118530 [-] on Build GRCh38 95
GenBankmRNA BE866300.1 - 1 - 2 98 Chr. 7 - 113117180 - 113118538 [-] on Build GRCh38 95
GenBankmRNA BG527392.1 - 1 - 2 115 Chr. 7 - 113117103 - 113118554 [-] on Build GRCh38 95
GenBankmRNA BG776999.1 - 1 - 2 116 Chr. 7 - 113117120 - 113118555 [-] on Build GRCh38 95
GenBankmRNA BG687464.1 - 1 - 2 115 Chr. 7 - 113117091 - 113118554 [-] on Build GRCh38 95
GenBankmRNA BG403932.1 - 1 - 2 Chr. 7 - 113117103 - 113118519 [-] on Build GRCh38 94
GenBankmRNA BG532857.1 - 1 - 2 115 Chr. 7 - 113117111 - 113118554 [-] on Build GRCh38 95
GenBankmRNA BG493956.1 - 1 - 2 118 Chr. 7 - 113117194 - 113118557 [-] on Build GRCh38 95
GenBankmRNA BI766819.1 - 1 - 2 97 Chr. 7 - 113116870 - 113118537 [-] on Build GRCh38 95
GenBankmRNA BM857697.1 - 1 - 2 86 Chr. 7 - 113117417 - 113118519 [-] on Build GRCh38 95
GenBankmRNA BG530236.1 - 1 - 2 115 Chr. 7 - 113117016 - 113118554 [-] on Build GRCh38 95
GenBankmRNA BP421775.1 - 1 - 2 77 Chr. 7 - 113117268 - 113118517 [-] on Build GRCh38 95
GenBankmRNA BP421774.1 - 1 - 2 Chr. 7 - 113117279 - 113118517 [-] on Build GRCh38 96
GenBankmRNA BG532952.1 - 1 - 2 115 Chr. 7 - 113117052 - 113118554 [-] on Build GRCh38 95
GenBankmRNA AW967865.1 - 1 - 2 88 Chr. 7 - 113117261 - 113118528 [-] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein-coupled receptor 85 GPR85 SREB,
SREB2
-30.3kb Sense Chr. 7 - 113080413 - 113087778 on NCBI Build GRCh38
Downstream protein phosphatase 1 regulatory subunit 3A PPP1R3A GM,
PP1G,
PPP1R3
758.2kb Sense Chr. 7 - 113876827 - 113919094 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012301.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001768.hg.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027682.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027682.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027682.hg.1 NR_024412.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027682.hg.1 TC07001768.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000397764.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000439551.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_015442.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay TC0700012301.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 ENST00000413744.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 ENST00000441359.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 NR_024412.1 Refseq
Affymetrix Human Clariom D Assay PSR0700188742.hg.1 TC0700012301.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline