This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03681035_m1
See other GSTM3 GE Assays ›
Gene Symbol:
GSTM3
Gene Name:
glutathione S-transferase mu 3
Gene Aliases:
GST5, GSTB, GSTM3-3, GTM3
Chromosome Location:
Chr.1: 109733932 - 109741038 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03681035_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024537.1 2 - 3 359 79
GenBank mRNA AK304300.1 - 1 - 2 87 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ051402.1 - 1 - 2 59 Chr. 1 - 109736975 - 109740297 [-] on Build GRCh38 79
GenBankmRNA BX343662.2 - 6 - 7 Chr. 1 - 109736964 - 109740293 [+] on Build GRCh38 79
GenBankmRNA DC417764.1 - 1 - 2 87 Chr. 1 - 109737140 - 109740325 [-] on Build GRCh38 79
GenBankmRNA BM792995.1 - 1 - 2 60 Chr. 1 - 109737114 - 109740298 [-] on Build GRCh38 79
GenBankmRNA BM793887.1 - 1 - 2 57 Chr. 1 - 109737069 - 109740295 [-] on Build GRCh38 79
GenBankmRNA CD638951.1 - 1 - 2 81 Chr. 1 - 109736994 - 109740316 [-] on Build GRCh38 79
GenBankmRNA BX343663.2 - 1 - 2 55 Chr. 1 - 109736926 - 109740293 [-] on Build GRCh38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glutathione S-transferase mu 5 GSTM5 GSTM5-5,
GTM5
-21.2kb Anti-sense Chr. 1 - 109711751 - 109718268 on NCBI Build GRCh38
Downstream EPS8 like 3 EPS8L3 EPS8R3 9.6kb Sense Chr. 1 - 109750080 - 109764022 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015211.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002962.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK304300.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
establishment of blood-nerve barrier
nitrobenzene metabolic process
xenobiotic catabolic process
response to estrogen
cellular detoxification of nitrogen compound
glutathione derivative biosynthetic process
glutathione transferase activity
enzyme binding
identical protein binding
protein homodimerization activity
glutathione binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 TC01002962.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 ENST00000256594.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 ENST00000361066.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 ENST00000540225.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01046059.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213388.hg.1 TC0100015211.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0