This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03806487_s1
See other LOC391322 GE Assays ›
Gene Symbol:
LOC391322 GSTT1-AS1
Gene Name:
D-dopachrome tautomerase-like
GSTT1 antisense RNA 1
Gene Aliases:
-
-
Chromosome Location:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs03806487_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC391322
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144931.1 NP_001138403.1 2 339 95
Gene Symbol: GSTT1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BF511344.1 - 2 95
BQ771818.1 - 2 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000143.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

decarboxylase

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human CNV Microarray 2x400K 271477 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3379341 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1953 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1954 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15505 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3379341 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3379341 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122937 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122949 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267977 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444720 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444729 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122943 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444723 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267950 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444716 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444726 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122943 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444716 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267977 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17267950 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122937 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444726 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444723 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444729 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122949 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32122930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11444720 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444768 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444722 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444770 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444745 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444761 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444727 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444715 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444758 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444728 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17267950 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444764 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444716 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17267980 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444763 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444720 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444756 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444744 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444760 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444723 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444749 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17267979 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444747 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444729 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444731 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444754 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444752 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444771 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P11444725 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P17267992 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8071808 - -
Affymetrix Human Genome U219 Array 11740968_x_at - -
Affymetrix Human Genome U219 Array 11740966_a_at - -
Affymetrix Human Genome U219 Array 11740967_PM_at - -
Affymetrix Human Genome U219 Array 11740966_PM_a_at - -
Affymetrix Human Genome U219 Array 11740968_PM_x_at - -
Affymetrix Human Genome U219 Array 11740967_at - -
Affymetrix Human Transcriptome Array 2.0 PSR22002595.hg.1 NM_001144931.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22002595.hg.1 TC22000143.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22002596.hg.1 NM_001144931.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22002596.hg.1 TC22000143.hg.1 HTA TC
Human Genome U95 Set 372_f_at - -
Human Genome U95 Set 372_PM_f_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A020220 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A020220 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 135595 - -
ABI Human Genome Survey Microarray Version 2.0 154739 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1798664 - -
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1798664 - -
Illumina human-6 v2.0 expression beadchip ILMN_1798664 - -
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1798664 - -
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1798664 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0