This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03829159_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs03829159_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ575803.1 - 1 Chr. 1 - 40298531 - 40299173 [+] on Build GRCh38 70
GenBankmRNA CA419063.1 - 1 Chr. 1 - 40298531 - 40299194 [+] on Build GRCh38 70
GenBankmRNA AA488854.1 - 1 Chr. 1 - 40298537 - 40298844 [+] on Build GRCh38 70
GenBankmRNA AW080015.1 - 1 Chr. 1 - 40298546 - 40298980 [+] on Build GRCh38 70
GenBankmRNA AW292156.1 - 1 Chr. 1 - 40298532 - 40299036 [+] on Build GRCh38 70
GenBankmRNA AK021682.1 - 1 1350 Chr. 1 - 40298549 - 40300029 [-] on Build GRCh38 70
GenBankmRNA AU145185.1 - 1 Chr. 1 - 40298549 - 40299111 [+] on Build GRCh38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc metallopeptidase STE24 ZMPSTE24 FACE-1,
FACE1,
HGPS,
PRO1,
STE24,
Ste24p
-4.4kb Anti-sense Chr. 1 - 40258050 - 40294184 on NCBI Build GRCh38
Downstream collagen type IX alpha 2 COL9A2 DJ39G22.4,
EDM2,
MED,
STL5
458.0 Sense Chr. 1 - 40300487 - 40317653 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline