This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03830787_s1
See other METTL9 GE Assays ›
Gene Symbol:
METTL9
Gene Name:
methyltransferase like 9
Gene Aliases:
CGI-81, DREV, DREV1, PAP1
Chromosome Location:
Chr.16: 21597221 - 21657473 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03830787_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL359211.1 - 1 351 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX108378.1 - 1 623 Chr. 16 - 21626504 - 21627240 [+] on Build GRCh38 71
GenBankmRNA AW451874.1 - 1 Chr. 16 - 21626948 - 21627411 [-] on Build GRCh38 71
GenBankmRNA AA630132.1 - 1 Chr. 16 - 21626949 - 21627392 [-] on Build GRCh38 71
GenBankmRNA AA157375.1 - 1 Chr. 16 - 21626972 - 21627393 [-] on Build GRCh38 71
GenBankmRNA AI536637.1 - 1 Chr. 16 - 21626909 - 21627394 [-] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101927814 LOC101927814 -27.4kb Anti-sense Chr. 16 - 21589698 - 21599385 on NCBI Build GRCh38
Downstream immunoglobulin superfamily member 6 IGSF6 DORA 13.9kb Anti-sense Chr. 16 - 21641284 - 21652666 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600007170.hg.1 ENST00000564271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007170.hg.1 ENST00000564271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007170.hg.1 ENST00000564271.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007170.hg.1 ENST00000564271.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007170.hg.1 ENST00000564271.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007170.hg.1 ENST00000564271.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline