Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | histone deacetylase 9 | HDAC9 |
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR |
-93.6kb | Anti-sense | Chr. 7 - 18086942 - 19003745 on NCBI Build GRCh38 |
| Downstream | twist family bHLH transcription factor 1 | TWIST1 |
ACS3, BPES2, BPES3, CRS, CRS1, CSO, SCS, TWIST, bHLHa38 |
19.7kb | Sense | Chr. 7 - 19113047 - 19117672 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map