This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03833107_s1
See other ABCG1 GE Assays ›
Gene Symbol:
ABCG1
Gene Name:
ATP binding cassette subfamily G member 1
Gene Aliases:
ABC8, WHITE1
Chromosome Location:
Chr.21: 42199689 - 42304389 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03833107_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011529806.1 XP_011528108.1 15 10037 64
XM_011529807.2 XP_011528109.1 15 10059 64
GenBank mRNA AF086547.1 - 1 146 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW449035.1 - 1 Chr. 21 - 42303993 - 42304389 [-] on Build GRCh38 64
GenBankmRNA BE550262.1 - 1 Chr. 21 - 42303833 - 42304387 [-] on Build GRCh38 64
GenBankmRNA AI457531.1 - 1 Chr. 21 - 42303916 - 42304387 [-] on Build GRCh38 64
GenBankmRNA AI818064.1 - 1 Chr. 21 - 42303907 - 42304389 [-] on Build GRCh38 64
GenBankmRNA BU742664.1 - 1 Chr. 21 - 42303880 - 42304389 [-] on Build GRCh38 64
GenBankmRNA AI167798.1 - 1 Chr. 21 - 42304016 - 42304389 [-] on Build GRCh38 64
GenBankmRNA BE674309.1 - 1 Chr. 21 - 42303821 - 42304389 [-] on Build GRCh38 64
GenBankmRNA AI150381.1 - 1 Chr. 21 - 42304040 - 42304389 [-] on Build GRCh38 64
GenBankmRNA W94442.1 - 146 Chr. - - [-] on Build 64
GenBankmRNA CK299542.1 - 1 Chr. 21 - 42303645 - 42304389 [-] on Build GRCh38 64
GenBankmRNA BF433594.1 - 1 Chr. 21 - 42303907 - 42304389 [-] on Build GRCh38 64
GenBankmRNA DB304723.1 - 1 Chr. 21 - 42303833 - 42304388 [-] on Build GRCh38 64
GenBankmRNA DB325086.1 - 1 Chr. 21 - 42303853 - 42304388 [-] on Build GRCh38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uromodulin like 1 UMODL1 -161.1kb Sense Chr. 21 - 42062959 - 42142996 on NCBI Build GRCh38
Downstream trefoil factor 3 TFF3 ITF,
P1B,
TFI
7.3kb Anti-sense Chr. 21 - 42311667 - 42315596 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
cholesterol metabolic process
detection of hormone stimulus
response to organic substance
negative regulation of macrophage derived foam cell differentiation
regulation of cholesterol esterification
positive regulation of cholesterol efflux
negative regulation of cholesterol storage
intracellular cholesterol transport
cholesterol efflux
phospholipid efflux
response to lipid
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
glycoprotein transport
lipoprotein metabolic process
cholesterol homeostasis
amyloid precursor protein catabolic process
reverse cholesterol transport
positive regulation of cholesterol biosynthetic process
transmembrane transport
phospholipid homeostasis
response to high density lipoprotein particle
toxin transport
ATP binding
phospholipid binding
phospholipid transporter activity
cholesterol binding
cholesterol transporter activity
toxin transporter activity
sterol-transporting ATPase activity
glycoprotein transporter activity
protein homodimerization activity
ADP binding
protein heterodimerization activity
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0