This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03835879_s1
See other CPSF4 GE Assays ›
Gene Symbol:
CPSF4
Gene Name:
cleavage and polyadenylation specific factor 4
Gene Aliases:
CPSF30, NAR, NEB-1, NEB1
Chromosome Location:
Chr.7: 99438920 - 99457377 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03835879_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPSF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017011700.1 XP_016867189.1 7 2421 80
XM_017011701.1 XP_016867190.1 7 2406 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA076703.1 - 1 149 Chr. 7 - 99455568 - 99455768 [+] on Build GRCh38 80
GenBankmRNA CR991793.1 - 1 451 Chr. 7 - 99455266 - 99456004 [+] on Build GRCh38 80
GenBankmRNA G30756.1 - 1 149 Chr. 7 - 99455568 - 99455768 [+] on Build GRCh38 80
GenBankmRNA BF527567.1 - 1 166 Chr. 7 - 99455551 - 99456200 [+] on Build GRCh38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATP5J2-PTCD1 readthrough ATP5J2-PTCD1 Anti-sense Chr. 7 - 99416739 - 99466201 on NCBI Build GRCh38
Downstream ATP synthase,
H+ transporting,
mitochondrial Fo complex subunit F2
ATP5J2 ATP5JL 2.4kb Anti-sense Chr. 7 - 99458161 - 99466201 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

tRNA splicing, via endonucleolytic cleavage and ligation
mRNA processing
modification by virus of host mRNA processing
protein binding
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0