This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03843043_s1
See other PSMG4 GE Assays ›
Gene Symbol:
PSMG4
Gene Name:
proteasome assembly chaperone 4
Gene Aliases:
C6orf86, PAC4, bA506K6.2
Chromosome Location:
Chr.6: 3254325 - 3308224 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03843043_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PSMG4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011514593.2 XP_011512895.1 3 60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY798448.2 - 1 460 Chr. 6 - 3303999 - 3304599 [-] on Build GRCh38 60
GenBankmRNA BQ773789.1 - 1 240 Chr. 6 - 3303724 - 3304361 [-] on Build GRCh38 60
GenBankmRNA AL833424.1 - 1 854 Chr. 6 - 3302440 - 3304993 [-] on Build GRCh38 60
GenBankmRNA FN068639.1 - 1 Chr. 6 - 3304074 - 3304177 [+] on Build GRCh38 60
GenBankmRNA BE671071.1 - 1 221 Chr. 6 - 3303803 - 3304357 [-] on Build GRCh38 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 22 member 23 SLC22A23 C6orf85 Sense Chr. 6 - 3268973 - 3456574 on NCBI Build GRCh38
Downstream uncharacterized LOC643327 LOC643327 153.7kb Anti-sense Chr. 6 - 3458703 - 3469212 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s)

proteasome assembly

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0