This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03847753_m1
See other LOC101929694 GE Assays ›
Gene Symbol:
LOC101929694
Gene Name:
uncharacterized LOC101929694
Gene Aliases:
-
Chromosome Location:
Chr.3: 112736449 - 112749319 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03847753_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101929694
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110017.1 3 - 4 1626 107
GenBank mRNA BC033333.1 - 3 - 4 1628 107

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 80 CCDC80 DRO1,
SSG1,
URB,
okuribin
-97.5kb Sense Chr. 3 - 112604386 - 112641143 on NCBI Build GRCh38
Downstream CD200 receptor 1 like CD200R1L CD200R2,
CD200RLa
71.2kb Sense Chr. 3 - 112814925 - 112847004 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline