This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03847869_s1
See other NSUN6 GE Assays ›
Gene Symbol:
NSUN6
Gene Name:
NOP2/Sun RNA methyltransferase family member 6
Gene Aliases:
4933414E04Rik, ARL5B-AS1, NOPD1
Chromosome Location:
Chr.10: 18512582 - 18659264 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03847869_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NSUN6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011519384.1 XP_011517686.1 2 162 78
GenBank mRNA AL698095.1 - 2 157 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC033534.1 - 1 852 Chr. 10 - 18656826 - 18659238 [-] on Build GRCh38 78
GenBankmRNA CA334862.1 - 1 Chr. 10 - 18658177 - 18727072 [+] on Build GRCh38 78
GenBankmRNA DB031997.1 - 2 157 Chr. 10 - 18651395 - 18659238 [-] on Build GRCh38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream calcium voltage-gated channel auxiliary subunit beta 2 CACNB2 CACNLB2,
CAVB2,
MYSB
-115.1kb Anti-sense Chr. 10 - 18140560 - 18545881 on NCBI Build GRCh38
Downstream ADP ribosylation factor like GTPase 5B ARL5B ARL8 97.0 Anti-sense Chr. 10 - 18659335 - 18681639 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011519384.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

RNA methylation
tRNA methylation
tRNA binding
RNA methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000414939.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000444660.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000449529.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000607346.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012529.hg.1 ENST00000607346.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline