This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03848673_s1
See other MSI2 GE Assays ›
Gene Symbol:
MSI2
Gene Name:
musashi RNA binding protein 2
Gene Aliases:
MSI2H
Chromosome Location:
Chr.17: 57255851 - 57684689 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03848673_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC035768.1 - 1 3255 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream A-kinase anchoring protein 1 AKAP1 AKAP,
AKAP121,
AKAP149,
AKAP84,
D-AKAP1,
PPP1R43,
PRKA1,
SAKAP84,
TDRD17
-363.6kb Sense Chr. 17 - 57085192 - 57121349 on NCBI Build GRCh38
Downstream uncharacterized LOC101927557 LOC101927557 33.6kb Anti-sense Chr. 17 - 57522248 - 57523597 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011220.hg.1 ENST00000616595.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011220.hg.1 ENST00000616595.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0