This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03851557_s1
See other EIF2AK2 GE Assays ›
Gene Symbol:
EIF2AK2
Gene Name:
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases:
EIF2AK1, PKR, PPP1R83, PRKR
Chromosome Location:
Chr.2: 37099210 - 37157065 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03851557_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF2AK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011532987.2 XP_011531289.1 17 9702 86
XM_017004503.1 XP_016859992.1 16 9569 86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA147004.1 - 1 Chr. 2 - 37099212 - 37099669 [+] on Build GRCh38 86
GenBankmRNA CB158514.1 - 1 441 Chr. 2 - 37099378 - 37099925 [-] on Build GRCh38 86
GenBankmRNA AI037957.1 - 1 Chr. 2 - 37099210 - 37099684 [+] on Build GRCh38 86
GenBankmRNA BG283489.1 - 1 443 Chr. 2 - 37099210 - 37099928 [-] on Build GRCh38 86
GenBankmRNA AI369437.1 - 1 Chr. 2 - 37099210 - 37099653 [+] on Build GRCh38 86
GenBankmRNA BQ182397.1 - 1 Chr. 2 - 37099210 - 37099715 [+] on Build GRCh38 86
GenBankmRNA CN404394.1 - 1 707 Chr. 2 - 37099411 - 37100176 [-] on Build GRCh38 86
GenBankmRNA AI241379.1 - 1 Chr. 2 - 37099211 - 37099664 [+] on Build GRCh38 86
GenBankmRNA D81020.1 - 1 69 Chr. 2 - 37099340 - 37099552 [-] on Build GRCh38 86
GenBankmRNA AI927369.1 - 1 Chr. 2 - 37099212 - 37099659 [+] on Build GRCh38 86
GenBankmRNA AW612409.1 - 1 Chr. 2 - 37099210 - 37099633 [+] on Build GRCh38 86
GenBankmRNA AI356183.1 - 1 Chr. 2 - 37099210 - 37099649 [+] on Build GRCh38 86
GenBankmRNA AA700984.1 - 1 Chr. 2 - 37099219 - 37099539 [+] on Build GRCh38 86
GenBankmRNA CA445747.1 - 1 Chr. 2 - 37099210 - 37099927 [+] on Build GRCh38 86
GenBankmRNA BE501096.1 - 1 Chr. 2 - 37099210 - 37099652 [+] on Build GRCh38 86
GenBankmRNA BU618209.1 - 1 Chr. 2 - 37099210 - 37099951 [+] on Build GRCh38 86
GenBankmRNA BQ029954.1 - 1 Chr. 2 - 37099210 - 37099693 [+] on Build GRCh38 86
GenBankmRNA BX362627.2 - 1 Chr. 2 - 37099241 - 37100292 [+] on Build GRCh38 86
GenBankmRNA AI371075.1 - 1 Chr. 2 - 37099210 - 37099632 [+] on Build GRCh38 86
GenBankmRNA CA448115.1 - 1 Chr. 2 - 37099210 - 37099996 [+] on Build GRCh38 86
GenBankmRNA BM684728.1 - 1 Chr. 2 - 37099210 - 37099957 [+] on Build GRCh38 86
GenBankmRNA AW874364.1 - 1 Chr. 2 - 37099210 - 37099676 [+] on Build GRCh38 86
GenBankmRNA BU677520.1 - 1 665 Chr. 2 - 37099421 - 37100146 [-] on Build GRCh38 86
GenBankmRNA N62286.1 - 1 Chr. 2 - 37099221 - 37099676 [+] on Build GRCh38 86
GenBankmRNA BP398333.1 - 1 228 Chr. 2 - 37099212 - 37099711 [-] on Build GRCh38 86
GenBankmRNA AL047596.1 - 1 Chr. 2 - 37099210 - 37099905 [+] on Build GRCh38 86
GenBankmRNA BQ002126.1 - 1 Chr. 2 - 37099210 - 37099731 [+] on Build GRCh38 86
GenBankmRNA DB297791.1 - 1 Chr. 2 - 37099212 - 37099762 [+] on Build GRCh38 86
GenBankmRNA AA236576.1 - 1 Chr. 2 - 37099212 - 37099644 [+] on Build GRCh38 85
GenBankmRNA AI686711.1 - 1 Chr. 2 - 37099210 - 37099632 [+] on Build GRCh38 86
GenBankmRNA BU507028.1 - 1 676 Chr. 2 - 37099270 - 37100160 [-] on Build GRCh38 86
GenBankmRNA CA448019.1 - 1 Chr. 2 - 37099210 - 37099996 [+] on Build GRCh38 86
GenBankmRNA AI670731.1 - 1 Chr. 2 - 37099211 - 37099696 [+] on Build GRCh38 86
GenBankmRNA D79312.1 - 1 159 Chr. 2 - 37099314 - 37099642 [-] on Build GRCh38 86
GenBankmRNA AA167012.1 - 1 Chr. 2 - 37099239 - 37099810 [+] on Build GRCh38 86
GenBankmRNA DB314025.1 - 1 Chr. 2 - 37099212 - 37099743 [+] on Build GRCh38 86
GenBankmRNA AA393792.1 - 1 Chr. 2 - 37099210 - 37099725 [+] on Build GRCh38 86
GenBankmRNA AI038137.1 - 1 Chr. 2 - 37099210 - 37099664 [+] on Build GRCh38 86
GenBankmRNA AA781508.1 - 1 Chr. 2 - 37099212 - 37099528 [+] on Build GRCh38 86
GenBankmRNA BU564286.1 - 1 88 Chr. 2 - 37099212 - 37099569 [-] on Build GRCh38 86
GenBankmRNA BU615395.1 - 1 Chr. 2 - 37099210 - 37099928 [+] on Build GRCh38 86
GenBankmRNA BM670045.1 - 1 Chr. 2 - 37099210 - 37099855 [+] on Build GRCh38 86
GenBankmRNA AI076122.1 - 1 Chr. 2 - 37099218 - 37099625 [+] on Build GRCh38 86
GenBankmRNA AI669226.1 - 1 Chr. 2 - 37099212 - 37099659 [+] on Build GRCh38 86
GenBankmRNA AI183289.1 - 1 Chr. 2 - 37099210 - 37099632 [+] on Build GRCh38 86
GenBankmRNA CR603904.1 - 1 2652 Chr. 2 - 37099241 - 37102135 [-] on Build GRCh38 86
GenBankmRNA BU633248.1 - 1 Chr. 2 - 37099210 - 37099973 [+] on Build GRCh38 86
GenBankmRNA BM930091.1 - 1 536 Chr. 2 - 37099366 - 37100019 [-] on Build GRCh38 86
GenBankmRNA AA477717.1 - 1 Chr. 2 - 37099212 - 37099621 [+] on Build GRCh38 86
GenBankmRNA AA635190.1 - 1 Chr. 2 - 37099210 - 37099533 [+] on Build GRCh38 86
GenBankmRNA BQ004466.1 - 1 Chr. 2 - 37099210 - 37099925 [+] on Build GRCh38 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G-patch domain containing 11 GPATCH11 CCDC75,
CENP-Y,
CENPY
Anti-sense Chr. 2 - 37084451 - 37099244 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKK activity
positive regulation of cytokine production
transcription, DNA-templated
translation
protein phosphorylation
negative regulation of cell proliferation
response to mechanical stimulus
response to virus
response to toxic substance
regulation of translational initiation by eIF2 alpha phosphorylation
negative regulation of translation
peptidyl-tyrosine phosphorylation
evasion or tolerance by virus of host immune response
endoplasmic reticulum unfolded protein response
response to lipopolysaccharide
positive regulation of chemokine production
positive regulation of stress-activated MAPK cascade
response to vitamin E
negative regulation of osteoblast proliferation
regulation of protein phosphatase type 2A activity
cellular response to amino acid starvation
response to interferon-alpha
positive regulation of apoptotic process
negative regulation of apoptotic process
response to exogenous dsRNA
negative regulation of viral genome replication
innate immune response
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
defense response to virus
regulation of NLRP3 inflammasome complex assembly
positive regulation of NIK/NF-kappaB signaling
regulation of hematopoietic progenitor cell differentiation
regulation of hematopoietic stem cell proliferation
regulation of hematopoietic stem cell differentiation
double-stranded RNA binding
protein kinase activity
protein serine/threonine kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
non-membrane spanning protein tyrosine kinase activity
protein binding
ATP binding
protein phosphatase type 2A regulator activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000571970.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573042.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 ENST00000573268.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq
Affymetrix Human Clariom D Assay TC1600009219.hg.1 NR_039999.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-6zkmz:80/100.66.79.198:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline