This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03852571_s1
See other C6ORF163 GE Assays ›
Gene Symbol:
C6orf163
Gene Name:
chromosome 6 open reading frame 163
Gene Aliases:
-
Chromosome Location:
Chr.6: 87343080 - 87365553 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03852571_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C6orf163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006715372.2 XP_006715435.1 1 1251 102
GenBank mRNA AK057638.1 - 1 1251 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI140943.1 - 1 Chr. 6 - 87357882 - 87358315 [-] on Build GRCh38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream small integral membrane protein 8 SMIM8 C6orf162,
dJ102H19.2
-14.5kb Sense Chr. 6 - 87322588 - 87342328 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1590 LINC01590 C6orf164,
dJ102H19.4
38.5kb Sense Chr. 6 - 87397124 - 87399741 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006715372.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008730.hg.1 ENST00000623904.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008730.hg.1 ENST00000623904.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline