This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03854580_s1
See other NUDT16 GE Assays ›
Gene Symbol:
NUDT16
Gene Name:
nudix hydrolase 16
Gene Aliases:
-
Chromosome Location:
Chr.3: 131381671 - 131388830 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03854580_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171905.1 NP_001165376.1 4 3860 100
NM_152395.2 NP_689608.2 3 4034 100
NR_033268.1 2 4121 100
GenBank mRNA AK091353.1 - 1 450 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA486949.1 - 1 450 Chr. 3 - 131386298 - 131386853 [+] on Build GRCh38 100
GenBankmRNA AA774801.1 - 1 179 Chr. 3 - 131386570 - 131387053 [+] on Build GRCh38 100
GenBankmRNA BF114837.1 - 1 135 Chr. 3 - 131386613 - 131387100 [+] on Build GRCh38 100
GenBankmRNA BQ706802.1 - 1 536 Chr. 3 - 131386212 - 131387066 [+] on Build GRCh38 100
GenBankmRNA BQ934230.1 - 1 494 Chr. 3 - 131386264 - 131387150 [+] on Build GRCh38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC339874 LOC339874 -4.8kb Anti-sense Chr. 3 - 131325092 - 131381475 on NCBI Build GRCh38
Downstream mitochondrial ribosomal protein L3 MRPL3 COXPD9,
MRL3,
RPML3
73.8kb Anti-sense Chr. 3 - 131462201 - 131503016 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008830.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUDT16

Gene Ontology Categories:

Function(s) Process(es)

adenosine to inosine editing
mRNA catabolic process
proteolysis
positive regulation of cell proliferation
snoRNA catabolic process
dephosphorylation
nucleobase-containing small molecule catabolic process
dITP catabolic process
IDP catabolic process
positive regulation of cell cycle process
XDP catabolic process
negative regulation of rRNA processing
positive regulation of double-strand break repair
magnesium ion binding
mRNA binding
GTP binding
metalloexopeptidase activity
manganese ion binding
snoRNA binding
dITP diphosphatase activity
identical protein binding
protein homodimerization activity
m7G(5')pppN diphosphatase activity
cobalt ion binding
inosine-diphosphatase activity
dIDP diphosphatase activity
nucleotide phosphatase activity, acting on free nucleotides
XTP binding
ITP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay PSR0300163013.hg.1 TC0300008830.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000502852.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000521288.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 ENST00000537561.5 Ensembl
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171905.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_001171906.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NM_152395.2 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq
Affymetrix Human Clariom D Assay TC0300008830.hg.1 NR_033268.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline