This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03863639_m1
See other STARD4-AS1 GE Assays ›
Gene Symbol:
STARD4-AS1
Gene Name:
STARD4 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 111512226 - 111739726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03863639_m1
Size
Availability Made To Order
Catalog # 4426961
Price 416.00
Your Price
Online offer: 390.65
390.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD4-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040093.1 1 - 2 280 100
GenBank mRNA CR749489.1 - 1 - 2 280 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream StAR related lipid transfer domain containing 4 STARD4 Anti-sense Chr. 5 - 111496033 - 111512597 on NCBI Build GRCh38
Downstream neuronal regeneration related protein NREP C5orf13,
D4S114,
P311,
PRO1873,
PTZ17,
SEZ17
59.7kb Anti-sense Chr. 5 - 111729303 - 111976931 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008312.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000522.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05007305.hg.1 TC05000522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05007305.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007305.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05007306.hg.1 TC05000522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05007306.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007306.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05007307.hg.1 TC05000522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05007307.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007307.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156906.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156906.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156906.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156906.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156906.hg.1 TC0500008312.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156904.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156904.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156904.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156904.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156904.hg.1 TC0500008312.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500156905.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156905.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156905.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156905.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156905.hg.1 TC0500008312.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline