This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03863641_m1
See other STARD4-AS1 GE Assays ›
Gene Symbol:
STARD4-AS1
Gene Name:
STARD4 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 111512226 - 111739726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03863641_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD4-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040093.1 5 - 6 1042 88
GenBank mRNA CR749489.1 - 5 - 6 1042 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream StAR related lipid transfer domain containing 4 STARD4 -217.0kb Anti-sense Chr. 5 - 111496033 - 111512597 on NCBI Build GRCh38
Downstream NREP antisense RNA 1 NREP-AS1 177.7kb Sense Chr. 5 - 111912508 - 112017306 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008312.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000522.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05007311.hg.1 TC05000522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05007311.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007311.hg.1 NR_040093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05007312.hg.1 TC05000522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05007312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05007312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156912.hg.1 TC0500008312.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500156914.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156914.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156914.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156914.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156914.hg.1 TC0500008312.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay PSR0500156913.hg.1 TC0500008312.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000500779.2 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 ENST00000513221.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq
Affymetrix Human Clariom D Assay TC0500008312.hg.1 NR_040093.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-mnvx7:80/100.66.78.106:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0