This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03866448_s1
See other GPR161 GE Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03866448_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR161
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267609.1 NP_001254538.1 7 4979 89
NM_001267610.1 NP_001254539.1 7 4942 89
NM_001267611.1 NP_001254540.1 6 4529 89
NM_001267612.1 NP_001254541.1 6 4485 89
NM_001267613.1 NP_001254542.1 5 4378 89
NM_001267614.1 NP_001254543.1 5 4111 89
NM_153832.2 NP_722561.1 8 4936 89
XM_005245055.2 XP_005245112.1 7 4903 89
XM_005245056.2 XP_005245113.1 6 4510 89
XM_005245057.4 XP_005245114.1 6 4527 89
XM_006711251.1 XP_006711314.1 8 4949 89
XM_006711253.2 XP_006711316.2 6 4742 89
XM_011509370.1 XP_011507672.1 8 5247 89
XM_011509371.2 XP_011507673.1 8 4693 89
XM_011509372.1 XP_011507674.1 8 5025 89
XM_011509373.2 XP_011507675.1 8 4951 89
XM_011509375.2 XP_011507677.1 8 4737 89
XM_011509376.2 XP_011507678.1 7 4868 89
XM_011509377.1 XP_011507679.1 6 4824 89
XM_011509378.1 XP_011507680.1 6 4796 89
XM_017000858.1 XP_016856347.1 7 5396 89
XM_017000859.1 XP_016856348.1 6 5350 89
XM_017000860.1 XP_016856349.1 7 5444 89
XM_017000861.1 XP_016856350.1 8 5490 89
XM_017000862.1 XP_016856351.1 8 5073 89
GenBank mRNA AK095302.1 - 1 1851 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX478213.1 - 1 90 Chr. 1 - 168082308 - 168082908 [-] on Build GRCh38 89
GenBankmRNA DA505903.1 - 1 454 Chr. 1 - 168082710 - 168083273 [-] on Build GRCh38 89
GenBankmRNA DA746071.1 - 1 91 Chr. 1 - 168082356 - 168082910 [-] on Build GRCh38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DDB1 and CUL4 associated factor 6 DCAF6 1200006M05Rik,
ARCAP,
IQWD1,
MSTP055,
NRIP,
PC326
-5.0kb Anti-sense Chr. 1 - 167936247 - 168075843 on NCBI Build GRCh38
Downstream TOR signaling pathway regulator TIPRL TIP,
TIP41,
TIPRL1
94.2kb Anti-sense Chr. 1 - 168178845 - 168202114 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016322.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR161

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0