Assay Details
| Gene Symbol: DDX11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038927.2 | 2 - 3 | 770 | 112 | ||
| GenBank mRNA | AK096360.1 | - | 2 - 3 | 774 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetraspanin 11 | TSPAN11 | VSSW1971 | -4.3kb | Anti-sense | Chr. 12 - 30926433 - 31016502 on NCBI Build GRCh38 |
| Downstream | DEAD/H-box helicase 11 | DDX11 |
CHL1, CHLR1, KRG2, WABS |
12.8kb | Anti-sense | Chr. 12 - 31073574 - 31104799 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001357.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281432 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281467 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281429 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713176 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281492 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281474 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281437 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281460 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281442 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281383 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281449 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281414 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281436 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281417 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28713230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08281490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16545079 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | AB353947.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | AF319622.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | AF399931.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | AY090613.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | AY234788.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | BC104894.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | BC104920.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 170748 | NM_178559.4 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 129651 | AB011169.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 129651 | AF009301.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 129651 | BC039293.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 129651 | NM_005885.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 260540 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 87280 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 545987 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 523837 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 889119 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 420996 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 526141 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 777804 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 896655 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 380341 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 216781 | BC001515.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 216781 | BC001841.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 216781 | BC009319.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 116112 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 432157 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 115079 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 108245 | AK095025.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 108245 | BC022957.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 108245 | NM_020207.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 161717 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 161558 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 17149 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 406721 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 163454 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 306368 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 362928 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 186830 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 417281 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 368189 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 391194 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 382804 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 176808 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02597800 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P11397277 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02597800 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02597800 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19503970 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02597800 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 38110 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 16588 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 142574 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 63935 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503959 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11402897 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19503945 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02597822 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11397277 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02597797 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02597800 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11395715 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02597822 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02597800 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P11397277 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02597797 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000535870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000535870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017738.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017738.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12017738.hg.1 | TC12001357.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000535870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000535870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | ENST00000618041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010271.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177836.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177836.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177836.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177836.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177836.hg.1 | TC1200010271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200177835.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177835.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177835.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177835.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177835.hg.1 | TC1200010271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200177834.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177834.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177834.hg.1 | ENST00000500527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200177834.hg.1 | NR_038927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200177834.hg.1 | TC1200010271.hg.1 | Clariom D TC |
| Human Genome U95 Set | 84415_PM_at | - | - |
| Human Genome U95 Set | 84415_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047448 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047449 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047448 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047449 | - | - |

DDX11-AS1
Genomic Map