This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03869934_s1
See other LOC105378374 GE Assays ›
Gene Symbol:
LOC105378374
Gene Name:
uncharacterized LOC105378374
Gene Aliases:
-
Chromosome Location:
Chr.10: 78164902 - 78178689 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03869934_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105378374
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK097674.1 - 1 1855 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB338161.1 - 1 Chr. 10 - 78173193 - 78173754 [+] on Build GRCh38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein S24 RPS24 DBA3,
S24
-116.4kb Anti-sense Chr. 10 - 78033760 - 78056813 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 856 LINC00856 73.3kb Anti-sense Chr. 10 - 78248625 - 78252645 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline