This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03870321_s1
See other ARHGAP19-SLIT1 GE Assays ›
Gene Symbol:
ARHGAP19-SLIT1
Gene Name:
ARHGAP19-SLIT1 readthrough (NMD candidate)
Gene Aliases:
-
Chromosome Location:
Chr.10: 97153042 - 97292673 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03870321_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP19-SLIT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AA431797.1 - 1 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB343169.1 - 1 Chr. 10 - 97153045 - 97153566 [+] on Build GRCh38 84
GenBankmRNA BF980148.1 - 1 Chr. 10 - 97153117 - 97153689 [+] on Build GRCh38 85
GenBankmRNA AK097919.1 - 1 2250 Chr. 10 - 97153045 - 97155433 [-] on Build GRCh38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SLIT1 antisense RNA 1 SLIT1-AS1 -49.3kb Anti-sense Chr. 10 - 97102756 - 97103747 on NCBI Build GRCh38
Downstream Rho GTPase activating protein 19 ARHGAP19 66.7kb Sense Chr. 10 - 97222173 - 97292673 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0