This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03876991_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs03876991_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM926504.1 - 1 500 Chr. 19 - 41549524 - 41550521 [+] on Build GRCh38 67
GenBankmRNA BG534399.1 - 1 501 Chr. 19 - 41549518 - 41550148 [+] on Build GRCh38 67
GenBankmRNA CD691542.1 - 1 531 Chr. 19 - 41549518 - 41550144 [+] on Build GRCh38 67
GenBankmRNA BG570372.1 - 1 500 Chr. 19 - 41549518 - 41550167 [+] on Build GRCh38 67
GenBankmRNA BG484601.1 - 1 497 Chr. 19 - 41549518 - 41550295 [+] on Build GRCh38 67
GenBankmRNA AX775927.1 - 1 753 Chr. 19 - 41549264 - 41550473 [+] on Build GRCh38 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 1480 LINC01480 -12.4kb Sense Chr. 19 - 41535498 - 41536904 on NCBI Build GRCh38
Downstream uncharacterized LOC105372404 LOC105372404 50.1kb Sense Chr. 19 - 41598625 - 41605984 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0