Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 13 member C | FAM13C | FAM13C1 | -289.6kb | Anti-sense | Chr. 10 - 59245933 - 59363186 on NCBI Build GRCh38 |
| Downstream | myoregulin | MRLN |
LINC00948, M1, MLN, MUSER1 |
83.5kb | Anti-sense | Chr. 10 - 59736990 - 59753445 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map