This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03880287_m1
See other PTPRD-AS2 GE Assays ›
Gene Symbol:
PTPRD-AS2
Gene Name:
PTPRD antisense RNA 2 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.9: 10613206 - 10620420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03880287_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRD-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110696.1 1 - 2 218 103
GenBank mRNA BC045688.1 - 1 - 2 218 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI601152.1 - 1 - 2 223 Chr. 9 - 10613202 - 10618218 [+] on Build GRCh38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type D
PTPRD HPTP,
HPTPD,
HPTPDELTA,
PTPD,
RPTPDELTA
-483.0 Anti-sense Chr. 9 - 8314246 - 10612840 on NCBI Build GRCh38
Downstream histone-lysine N-methyltransferase SETMAR-like LOC105375975 993.6kb Anti-sense Chr. 9 - 11610222 - 11618749 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0