This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03881461_s1
See other GTF3C4 GE Assays ›
Gene Symbol:
GTF3C4
Gene Name:
general transcription factor IIIC subunit 4
Gene Aliases:
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
Chromosome Location:
Chr.9: 132670035 - 132694955 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03881461_s1
Size
Availability Made To Order
Catalog # 4426961
Price 416.00
Your Price
Online offer: 390.65
390.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF3C4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012204.3 NP_036336.2 5 6718 83
NR_133925.1 6 6756 83
XM_017015318.1 XP_016870807.1 5 4501 83
GenBank mRNA BX537622.1 - 1 1747 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN343543.1 - 1 583 Chr. 9 - 132692055 - 132692786 [+] on Build GRCh38 83
GenBankmRNA H07955.1 - 1 102 Chr. 9 - 132692529 - 132692994 [+] on Build GRCh38 83
GenBankmRNA R51777.1 - 101 Chr. - - [-] on Build 83
GenBankmRNA BM802903.1 - 1 210 Chr. 9 - 132692435 - 132693325 [+] on Build GRCh38 83
GenBankmRNA BQ428209.1 - 1 483 Chr. 9 - 132692148 - 132693010 [+] on Build GRCh38 83
GenBankmRNA BM475335.1 - 1 451 Chr. 9 - 132692180 - 132693032 [+] on Build GRCh38 83
GenBankmRNA BM129603.1 - 1 509 Chr. 9 - 132692122 - 132692707 [+] on Build GRCh38 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Downstream adenylate kinase 8 AK8 AK 8,
C9orf98,
DDX31
30.6kb Anti-sense Chr. 9 - 132725574 - 132878811 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF3C4

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
histone acetylation
5S class rRNA transcription from RNA polymerase III type 1 promoter
tRNA transcription from RNA polymerase III promoter
positive regulation of catalytic activity
DNA binding
histone acetyltransferase activity
protein binding
enzyme activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline