This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03885463_s1
See other MSRA GE Assays ›
Gene Symbol:
MSRA
Gene Name:
methionine sulfoxide reductase A
Gene Aliases:
PMSR
Chromosome Location:
Chr.8: 10054224 - 10428891 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03885463_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK123132.1 - 1 646 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 124-1 MIR124-1 MIR124A,
MIR124A1,
MIRN124-1,
MIRN124A1,
mir-124-1
-523.5kb Anti-sense Chr. 8 - 9903388 - 9903472 on NCBI Build GRCh38
Downstream uncharacterized LINCR-0001 LINCR-0001 45.7kb Sense Chr. 8 - 10474565 - 10481974 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline