This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03887962_s1
See other AUH GE Assays ›
Gene Symbol:
AUH
Gene Name:
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases:
-
Chromosome Location:
Chr.9: 91213815 - 91361969 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03887962_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AUH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK124142.1 - 1 3951 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB342426.1 - 1 Chr. 9 - 91215536 - 91216118 [+] on Build GRCh38 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 484 LINC00484 C9orf73 -32.8kb Anti-sense Chr. 9 - 91119062 - 91182762 on NCBI Build GRCh38
Downstream nuclear factor,
interleukin 3 regulated
NFIL3 E4BP4,
IL3BP1,
NF-IL3A,
NFIL3A
189.4kb Sense Chr. 9 - 91409045 - 91425063 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0