This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03891129_s1
See other NECTIN3 GE Assays ›
Gene Symbol:
NECTIN3
Gene Name:
nectin cell adhesion molecule 3
Gene Aliases:
CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3
Chromosome Location:
Chr.3: 111071759 - 111201444 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03891129_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NECTIN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017006123.1 XP_016861612.1 10 9366 95
XM_017006126.1 XP_016861615.1 9 9297 95
GenBank mRNA AK125090.1 - 1 1855 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA R81020.1 - 103 Chr. - - [-] on Build 95
GenBankmRNA BQ776010.1 - 1 Chr. 3 - 111200764 - 111201446 [-] on Build GRCh38 95
GenBankmRNA R80523.1 - 136 Chr. - - [-] on Build 95
GenBankmRNA D63117.1 - 1 40 Chr. 3 - 111200850 - 111201274 [+] on Build GRCh38 95
GenBankmRNA AA401672.1 - 1 197 Chr. 3 - 111200693 - 111200952 [+] on Build GRCh38 95
GenBankmRNA N39709.1 - 1 89 Chr. 3 - 111200800 - 111201156 [+] on Build GRCh38 95
GenBankmRNA D79920.1 - 1 40 Chr. 3 - 111200850 - 111201300 [+] on Build GRCh38 95
GenBankmRNA CA419745.1 - 1 Chr. 3 - 111200699 - 111201446 [-] on Build GRCh38 95
GenBankmRNA BG434674.1 - 1 343 Chr. 3 - 111200546 - 111201129 [+] on Build GRCh38 95
GenBankmRNA BM310697.1 - 1 435 Chr. 3 - 111200456 - 111200953 [+] on Build GRCh38 95
GenBankmRNA R82313.1 - 137 Chr. - - [-] on Build 95
GenBankmRNA BM674016.1 - 1 Chr. 3 - 111200779 - 111201446 [-] on Build GRCh38 96
GenBankmRNA BE767724.1 - 1 79 Chr. 3 - 111200815 - 111201230 [+] on Build GRCh38 95
GenBankmRNA CN479566.1 - 1 Chr. 3 - 111200721 - 111201446 [-] on Build GRCh38 95
GenBankmRNA BQ027069.1 - 1 Chr. 3 - 111200726 - 111201446 [-] on Build GRCh38 95
GenBankmRNA BE866923.1 - 481 Chr. - - [-] on Build 95
GenBankmRNA R24924.1 - 136 Chr. - - [-] on Build 95
GenBankmRNA CB305607.1 - 1 Chr. 3 - 111200713 - 111201446 [-] on Build GRCh38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PVRL3 antisense RNA 1 PVRL3-AS1 -129.1kb Anti-sense Chr. 3 - 111045315 - 111069959 on NCBI Build GRCh38
Downstream CD96 molecule CD96 TACTILE 340.6kb Sense Chr. 3 - 111542079 - 111665996 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

lens morphogenesis in camera-type eye
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
spermatid development
cell recognition
fertilization
single organismal cell-cell adhesion
adherens junction organization
retina morphogenesis in camera-type eye
receptor activity
receptor binding
protein binding
protein homodimerization activity
protein heterodimerization activity
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0