This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03892664_m1
See other FAM234B GE Assays ›
Gene Symbol:
FAM234B
Gene Name:
family with sequence similarity 234 member B
Gene Aliases:
KIAA1467
Chromosome Location:
Chr.12: 12927680 - 13083449 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03892664_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM234B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125676.1 - 2 - 3 134 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA245322.1 - 3 - 4 233 Chr. 12 - 13000433 - 13019866 [+] on Build GRCh38 71
GenBankmRNA DA243448.1 - 2 - 3 139 Chr. 12 - 13008094 - 13019966 [+] on Build GRCh38 71
GenBankmRNA DB001536.1 - 3 - 4 134 Chr. 12 - 13000430 - 13019927 [+] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream 5-hydroxytryptamine receptor 7 pseudogene 1 HTR7P1 HTR7P -3.3kb Sense Chr. 12 - 13000442 - 13004830 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000538670.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000538670.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000543321.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 ENST00000624664.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Clariom D Assay TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Clariom D Assay TC1200006904.hg.1 NR_002774.3 Refseq
Affymetrix Human Clariom D Assay TC1200006904.hg.1 NR_002774.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline