This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03897549_s1
See other AOAH GE Assays ›
Gene Symbol:
AOAH
Gene Name:
acyloxyacyl hydrolase
Gene Aliases:
-
Chromosome Location:
Chr.7: 36512042 - 36724549 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03897549_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AOAH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK130782.1 - 1 1442 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream anillin actin binding protein ANLN FSGS8,
Scraps,
scra
-84.2kb Anti-sense Chr. 7 - 36389803 - 36453791 on NCBI Build GRCh38
Downstream AOAH intronic transcript 1 AOAH-IT1 57.9kb Sense Chr. 7 - 36597834 - 36600120 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000431496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000431496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000567053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 ENST00000567053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000430589.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000431496.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000431496.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000562423.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563144.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000563202.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000564604.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000565892.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566178.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000566321.5 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000567053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 ENST00000567053.1 Ensembl
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_036475.2 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq
Affymetrix Human Clariom D Assay TC1500006681.hg.1 NR_037599.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline