This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03900699_s1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03900699_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011524342.2 XP_011522644.1 5 7214 88
XM_017024192.1 XP_016879681.1 5 7218 88
XM_017024194.1 XP_016879683.1 5 3367 88
XM_017024195.1 XP_016879684.1 5 3371 88
GenBank mRNA AK054819.1 - 1 1351 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CK824940.1 - 1 297 Chr. 17 - 54956801 - 54957372 [-] on Build GRCh38 88
GenBankmRNA DB296600.1 - 1 Chr. 17 - 54956760 - 54957309 [+] on Build GRCh38 88
GenBankmRNA BM272070.1 - 1 370 Chr. 17 - 54956877 - 54957435 [-] on Build GRCh38 88
GenBankmRNA BM724325.1 - 1 541 Chr. 17 - 54956814 - 54957617 [-] on Build GRCh38 88
GenBankmRNA BX105177.1 - 1 147 Chr. 17 - 54956754 - 54957223 [-] on Build GRCh38 88
GenBankmRNA BU741603.1 - 1 Chr. 17 - 54956750 - 54957426 [+] on Build GRCh38 88
GenBankmRNA AA995327.1 - 1 Chr. 17 - 54956757 - 54957202 [+] on Build GRCh38 88
GenBankmRNA AA826563.1 - 1 Chr. 17 - 54956752 - 54957164 [+] on Build GRCh38 88
GenBankmRNA BM929832.1 - 1 138 Chr. 17 - 54956769 - 54957213 [-] on Build GRCh38 88
GenBankmRNA AA909874.1 - 1 Chr. 17 - 54956750 - 54957171 [+] on Build GRCh38 88
GenBankmRNA BF087511.1 - 1 78 Chr. 17 - 54956845 - 54957143 [-] on Build GRCh38 88
GenBankmRNA CB110914.1 - 1 269 Chr. 17 - 54956821 - 54957344 [-] on Build GRCh38 88
GenBankmRNA DB315328.1 - 1 Chr. 17 - 54956760 - 54957311 [+] on Build GRCh38 88
GenBankmRNA AI972122.1 - 1 Chr. 17 - 54956796 - 54957163 [+] on Build GRCh38 88
GenBankmRNA W92974.1 - 283 Chr. - - [-] on Build 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kinesin family member 2B KIF2B -1.1mb Anti-sense Chr. 17 - 53822878 - 53825212 on NCBI Build GRCh38
Downstream syntaxin binding protein 4 STXBP4 Synip 10.3kb Anti-sense Chr. 17 - 54968728 - 55164285 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
respiratory chain complex IV assembly
aerobic respiration
negative regulation of glucokinase activity
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0