This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03902409_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs03902409_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI939308.1 - 1 Chr. 3 - 23923700 - 23924373 [-] on Build GRCh38 85
GenBankmRNA DB317633.1 - 1 Chr. 3 - 23923732 - 23924268 [-] on Build GRCh38 85
GenBankmRNA AI089885.1 - 1 Chr. 3 - 23923891 - 23924374 [-] on Build GRCh38 85
GenBankmRNA AK055501.1 - 1 1746 Chr. 3 - 23922255 - 23924373 [+] on Build GRCh38 85
GenBankmRNA DB317025.1 - 1 Chr. 3 - 23923832 - 23924373 [-] on Build GRCh38 85
GenBankmRNA BG112070.1 - 133 Chr. - - [-] on Build 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NFKB inhibitor interacting Ras like 1 NKIRAS1 KBRAS1,
kappaB-Ras1
-5.2kb Anti-sense Chr. 3 - 23889536 - 23917046 on NCBI Build GRCh38
Downstream nuclear receptor subfamily 1 group D member 2 NR1D2 BD73,
EAR-1R,
RVR
20.9kb Sense Chr. 3 - 23945260 - 23980618 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline