This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03902734_s1
See other CECR2 GE Assays ›
Gene Symbol:
CECR2
Gene Name:
CECR2, histone acetyl-lysine reader
Gene Aliases:
-
Chromosome Location:
Chr.22: 17359949 - 17558155 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03902734_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CECR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290046.1 NP_001276975.1 19 9102 74
NM_001290047.1 NP_001276976.1 19 9574 74
XM_006724079.3 XP_006724142.1 7 8017 74
XM_011546128.2 XP_011544430.1 18 9178 74
XM_011546129.2 XP_011544431.1 19 9178 74
XM_011546132.2 XP_011544434.1 19 9097 74
XM_017028785.1 XP_016884274.1 19 9259 74
GenBank mRNA AK055696.1 - 1 1049 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ950712.1 - 1 232 Chr. 22 - 17557370 - 17558142 [+] on Build GRCh38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cat eye syndrome chromosome region,
candidate 3 (non-protein coding)
CECR3 -289.8kb Anti-sense Chr. 22 - 17256859 - 17266733 on NCBI Build GRCh38
Downstream solute carrier family 25 member 18 SLC25A18 GC2 5.0kb Sense Chr. 22 - 17562470 - 17590890 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cytokinesis
apoptotic DNA fragmentation
cytoskeleton organization
vesicle-mediated transport
covalent chromatin modification
neural tube development
ATP-dependent chromatin remodeling
execution phase of apoptosis

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0