This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03903183_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Human
Assay Type:
Non-coding
Assay ID Hs03903183_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW890029.1 - 1 Chr. 17 - 5113753 - 5114332 [-] on Build GRCh38 91
GenBankmRNA AK056005.1 - 1 650 Chr. 17 - 5113359 - 5115004 [+] on Build GRCh38 91
GenBankmRNA AI693543.1 - 1 Chr. 17 - 5113884 - 5114381 [-] on Build GRCh38 91
GenBankmRNA AW899455.1 - 1 Chr. 17 - 5113718 - 5114275 [-] on Build GRCh38 91
GenBankmRNA AI809361.1 - Chr. - - [-] on Build 91
GenBankmRNA AW890131.1 - 1 Chr. 17 - 5113718 - 5114275 [-] on Build GRCh38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101928000 LOC101928000 Sense Chr. 17 - 5111935 - 5114382 on NCBI Build GRCh38
Downstream ubiquitin specific peptidase 6 USP6 HRP1,
TRE17,
TRE2,
TRESMCR,
Tre-2,
USP6-short
1.4kb Sense Chr. 17 - 5116438 - 5175032 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline