This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03907351_s1
See other NXPE1 GE Assays ›
Gene Symbol:
NXPE1
Gene Name:
neurexophilin and PC-esterase domain family member 1
Gene Aliases:
FAM55A
Chromosome Location:
Chr.11: 114518946 - 114560244 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03907351_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011542594.2 XP_011540896.1 8 5099 100
XM_011542595.2 XP_011540897.1 9 5211 100
XM_011542596.2 XP_011540898.1 9 5452 100
XM_011542597.2 XP_011540899.1 8 5340 100
XM_011542598.2 XP_011540900.1 7 5040 100
XM_011542599.2 XP_011540901.1 7 5281 100
GenBank mRNA AK127883.1 - 1 1949 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RNA exonuclease 2 REXO2 CGI-114,
REX2,
RFN,
SFN
-68.7kb Anti-sense Chr. 11 - 114439386 - 114450279 on NCBI Build GRCh38
Downstream neurexophilin and PC-esterase domain family member 4 NXPE4 C11orf33,
FAM55D
49.6kb Sense Chr. 11 - 114570591 - 114596215 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0