Assay Details
| Gene Symbol: RBM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF116667.2 | - | 3 | 698 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 331 | LINC00331 | NCRNA00331 | -536.4kb | Sense | Chr. 13 - 78787319 - 78840050 on NCBI Build GRCh38 |
| Downstream | RBM26 antisense RNA 1 | RBM26-AS1 | 28.8kb | Anti-sense | Chr. 13 - 79406309 - 79424336 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RBM26
Genomic Map